Oxazolo[4,5-b]pyridin-2(3H)-one ≥95%

Supplier: Matrix Scientific

Synonyms: 2,3-Dihydropyrido[2,3-d]oxazol-2-one

073262-1G 073262-5G 073262-10G
102529-244EA 55.38 USD
102529-244 102529-246 102529-248
Oxazolo[4,5-b]pyridin-2(3H)-one ≥95%
Oxazolo[4,5-b]pyridin-2(3H)-one

Hazard: Irritant

Formula: C₆H₄N₂O₂
MW: 136.11 g/mol
MDL Number: MFCD00204215
CAS Number: 60832-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR