5-Nitropyridine-2-carbonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Cyano-5-nitropyridine, 5-Nitropicolinonitrile

073261-1G 073261-5G
102529-240EA 126.15 USD
102529-240 102529-242
5-Nitropyridine-2-carbonitrile ≥95%
5-Nitropyridine-2-carbonitrile

Hazard: Irritant

Formula: C₆H₃N₃O₂
MW: 149.1069 g/mol
MDL Number: MFCD07368184
CAS Number: 100367-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR