5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine ≥95%

Supplier: Matrix Scientific

073260-1G 073260-5G
102529-234EA 364.62 USD
102529-234 102529-236
5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine ≥95%
5-Bromo-2-(2,5-dimethyl-1H-pyrrol-1-yl)pyridine

Hazard: Irritant

Formula: C₁₁H₁₁BrN₂
MW: 251.1200 g/mol
MDL Number: MFCD01125286
CAS Number: 228710-82-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR