5-Bromo-4-chloro-6-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073259-250MG 073259-1G
102529-226EA 846.15 USD
102529-226 102529-228
5-Bromo-4-chloro-6-methylpyrimidine ≥95%
5-Bromo-4-chloro-6-methylpyrimidine

Hazard: Irritant

Formula: C₅H₄BrClN₂
MW: 207.46 g/mol
MDL Number: MFCD00234032
CAS Number: 3438-55-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR