Ethyl-4-bromo-1H-indole-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Ethyl 4-bromo-1H-indole-2-carboxylate

073258-1G 073258-5G
102529-222EA 407.69 USD
102529-222 102529-224
Ethyl-4-bromo-1H-indole-2-carboxylate ≥95%
Ethyl-4-bromo-1H-indole-2-carboxylate

Hazard: Irritant

Formula: C₁₁H₁₀BrNO₂
MW: 268.1066 g/mol
MDL Number: MFCD12174997
CAS Number: 103858-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR