Ethyl-7-bromo-4-hydroxyquinoline-3-carboxylate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 7-Bromo-4-hydroxy-3-quinolinecarboxylate
073257-1G
073257-5G
102529-216EA
640
USD
102529-216
102529-218
Ethyl-7-bromo-4-hydroxyquinoline-3-carboxylate ≥95%
Ethyl 7-bromo-4-hydroxyquinoline-3-carboxylate
Hazard: Irritant
Formula:
C₁₂H₁₀BrNO₃ MW: 296.1200 g/mol |
MDL Number:
MFCD00487280 CAS Number: 179943-57-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...