4-(Dimethoxymethyl)-2-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073256-1G 073256-5G
102529-212EA 100 USD
102529-212 102529-214
4-(Dimethoxymethyl)-2-methylpyrimidine ≥95%
4-(Dimethoxymethyl)-2-methylpyrimidine

Hazard: Irritant

Formula: C₈H₁₂N₂O₂
MW: 168.19 g/mol
MDL Number: MFCD00085137
CAS Number: 175277-33-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR