tert-Butyl-5-nitro-1H-indole-1-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 5-nitro-1H-indole-1-carboxylate

073255-1G 073255-5G
102529-208EA 209.23 USD
102529-208 102529-210
tert-Butyl-5-nitro-1H-indole-1-carboxylate ≥95%
tert-Butyl-5-nitro-1H-indole-1-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₄
MW: 262.2600 g/mol
MDL Number: MFCD09953094
CAS Number: 166104-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR