N-Ethyl-3-nitropyridin-4-amine ≥95%

Supplier: Matrix Scientific

073253-1G 073253-5G
102529-198EA 304.62 USD
102529-198 102529-200
N-Ethyl-3-nitropyridin-4-amine ≥95%
N-Ethyl-3-nitropyridin-4-amine

Hazard: Irritant

Formula: C₇H₉N₃O₂
MDL Number: MFCD02266948
CAS Number: 562825-95-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR