Methyl-2-amino-5-methylnicotinate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 2-amino-5-methylnicotinate

073252-250MG 073252-1G 073252-5G
102529-190EA 640 USD
102529-190 102529-192 102529-194
Methyl-2-amino-5-methylnicotinate ≥95%
Methyl-2-amino-5-methylnicotinate

Hazard: Irritant

Formula: C₈H₁₀N₂O₂
MW: 166.1800 g/mol
MDL Number: MFCD07375375
CAS Number: 886371-10-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR