1-Boc-3-cyanopiperidine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-butyl 3-cyanopiperidine-1-carboxylate, 1-Boc-3-cyanopiperidine, 3-Cyano-1-piperidinecarboxylic acid tert-Butyl ester, tert-Butyl 3-Cyano-1-piperidinecarboxylate, 1-Boc-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-piperidinecarbonitrile, 1-tert-Butoxycarbonyl-3-cyanopiperidine

073251-1G 073251-5G 073251-25G
102529-184EA 15.38 USD
102529-184 102529-186 102833-616
1-Boc-3-cyanopiperidine ≥95%
1-Boc-3-cyanopiperidine

Hazard: Toxic/Moisture Sensitive

Formula: C₁₁H₁₈N₂O₂
MW: 210.27 g/mol
Melting Pt: 60…65 °C
MDL Number: MFCD01861222
CAS Number: 91419-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR