6-Aminopyridin-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Aminopyridin-3-ol, 2-Amino-5-hydroxypyridine

073250-1G 073250-5G
102529-180EA 253.85 USD
102529-180 102529-182
6-Aminopyridin-3-ol ≥95%
6-Aminopyridin-3-ol

Hazard: Irritant/Refrigerate

Formula: C₅H₆N₂O
MW: 110.1100 g/mol
MDL Number: MFCD03541056
CAS Number: 55717-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR