5-Aminopyridin-3-ol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Amino-5-hydroxypyridine

073244-250MG 073244-1G 073244-5G
102529-146EA 400 USD
102529-146 102529-148 102529-150
5-Aminopyridin-3-ol ≥95%
5-Aminopyridin-3-ol

Hazard: Irritant

Formula: C₅H₆N₂O
MW: 110.1139 g/mol
MDL Number: MFCD04114213
CAS Number: 3543-01-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR