4-Chloro-3-formylbenzonitrile ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-5-cyanobenzaldehyde

073242-1G 073242-5G
102529-136EA 135.38 USD
102529-136 102529-138
4-Chloro-3-formylbenzonitrile ≥95%
4-Chloro-3-formylbenzonitrile

Hazard: Irritant

Formula: C₈H₄ClNO
MW: 165.5800 g/mol
MDL Number: MFCD11035756
CAS Number: 105191-41-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR