1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride ≥95%

Supplier: Matrix Scientific

073241-250MG 073241-1G 073241-5G
102529-130EA 640 USD
102529-130 102529-132 102529-134
1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride ≥95%
1-Oxa-8-azaspiro[4.5]decan-2-one hydrochloride

Hazard: Irritant

Formula: C₈H₁₄ClNO₂
MW: 191.6600 g/mol
MDL Number: MFCD11869757
CAS Number: 374794-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR