N,N-Dimethyl-2-(piperazin-1-yl)benzamideoxalate ≥95%

Supplier: Matrix Scientific

073240-250MG 073240-1G 073240-5G
102529-124EA 640 USD
102529-124 102529-126 102529-128
N,N-Dimethyl-2-(piperazin-1-yl)benzamideoxalate ≥95%
N,N-Dimethyl-2-(piperazin-1-yl)benzamideoxalate

Hazard: Irritant

Formula: C₁₅H₂₁N₃O₅
CAS Number: 494782-69-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR