2-Chlorobenzo[d]thiazol-6-amine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Amino-2-chlorobenzothiazole

073239-1G 073239-5G 073239-10G
102529-118EA 569.23 USD
102529-118 102529-120 102529-122
2-Chlorobenzo[d]thiazol-6-amine ≥95%
2-Chlorobenzo[d]thiazol-6-amine

Hazard: Irritant

Formula: C₇H₅ClN₂S
MW: 184.646 g/mol
MDL Number: MFCD08276901
CAS Number: 2406-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR