2,4-Dimethoxy-5H-pyrrolo[3,2-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073238-250MG 073238-1G
102529-114EA 684.62 USD
102529-114 102529-116
2,4-Dimethoxy-5H-pyrrolo[3,2-d]pyrimidine ≥95%
2,4-Dimethoxy-5H-pyrrolo[3,2-d]pyrimidine

Hazard: Irritant

Formula: C₈H₉N₃O₂
CAS Number: 84538-40-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR