2-Bromo-5-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073235-250MG 073235-1G 073235-5G
102529-096EA 489.23 USD
102529-096 102529-098 102529-100
2-Bromo-5-methylpyrimidine ≥95%
2-Bromo-5-methylpyrimidine

Hazard: Irritant

Formula: C₅H₅BrN₂
MW: 173.01 g/mol
MDL Number: MFCD09753737
CAS Number: 150010-20-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR