6-Bromo-2-hydroxyquinoxaline ≥95%

Supplier: Matrix Scientific

073234-250MG 073234-1G 073234-5G
102529-090EA 400 USD
102529-090 102529-092 102529-094
6-Bromo-2-hydroxyquinoxaline ≥95%
6-Bromo-2-hydroxyquinoxaline

Hazard: Irritant

Formula: C₈H₅BrN₂O
MDL Number: MFCD09834132
CAS Number: 55687-34-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR