Benzo[b]thiophene-2-carboxylic acid ≥95%

Supplier: Matrix Scientific

073233-5G 073233-25G
102529-086EA 36.92 USD
102529-086 102663-942
Benzo[b]thiophene-2-carboxylic acid ≥95%
Benzo[b]thiophene-2-carboxylic acid

Hazard: Irritant

Formula: C₉H₆O₂S
MW: 178.22 g/mol
Melting Pt: 236-242°C
MDL Number: MFCD00051636
CAS Number: 6314-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR