1-Methylpyrrolidine-3-carboxylic acid hydrochloride ≥95%

Supplier: Matrix Scientific

073232-250MG 073232-1G 073232-5G
102529-078EA 355.38 USD
102529-078 102529-080 102529-082
1-Methylpyrrolidine-3-carboxylic acid hydrochloride ≥95%
1-Methylpyrrolidine-3-carboxylic acid hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClNO₂
MW: 165.6200 g/mol
MDL Number: MFCD06738921
CAS Number: 50585-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR