4-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one ≥95%
Supplier: Matrix Scientific
073231-250MG
073231-1G
102529-074EA
853.85
USD
102529-074
102529-076
4-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one ≥95%
4-Bromo-1H-pyrrolo[2,3-b]pyridin-2(3H)-one
Hazard: Irritant
Formula:
C₇H₅BrN₂O MW: 213.0314 g/mol |
MDL Number:
MFCD12406103 CAS Number: 1086064-49-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...