Methyl-4-methylthiazole-5-carboxylate ≥95%

Supplier: Matrix Scientific

073229-1G 073229-5G
102529-064EA 100 USD
102529-064 102529-066
Methyl-4-methylthiazole-5-carboxylate ≥95%
Methyl-4-methylthiazole-5-carboxylate

Hazard: Irritant

Formula: C₆H₇NO₂S
CAS Number: 81569-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR