3-Aminopyrazine-2-carbonitrile ≥95%
Supplier: Matrix Scientific
073226-1G
073226-5G
102529-050EA
640
USD
102529-050
102529-052
3-Aminopyrazine-2-carbonitrile ≥95%
3-Aminopyrazine-2-carbonitrile
Hazard: Irritant
Formula:
C5H4N4 MW: 120.1121 g/mol |
MDL Number:
MFCD02091510 CAS Number: 25911-65-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...