Isoquinoline-3-amine ≥95%
Supplier: Matrix Scientific
Synonyms:
Isoquinolin-3-amine, 3-Aminoisoquinoline
073224-1G
073224-5G
102529-040EA
249.23
USD
102529-040
102529-042
Isoquinoline-3-amine ≥95%
Isoquinoline-3-amine
Hazard: Irritant
Formula:
C₉H₈N₂ MW: 144.1700 g/mol |
MDL Number:
MFCD00102190 CAS Number: 25475-67-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...