2-Chloroquinoline ≥95%

Supplier: Matrix Scientific

073221-5G 073221-10G 073221-100G
102529-022EA 67.69 USD
102529-022 102529-024 102529-026
2-Chloroquinoline ≥95%
2-Chloroquinoline

Hazard: Irritant

Formula: C₉H₆ClN
MDL Number: MFCD00006741
CAS Number: 612-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR