5-Bromo-2-iodobenzaldehyde ≥95%

Supplier: Matrix Scientific

073220-1G 073220-5G
102529-018EA 533.85 USD
102529-018 102529-020
5-Bromo-2-iodobenzaldehyde ≥95%
5-Bromo-2-iodobenzaldehyde

Hazard: Irritant

Formula: C₇H₄BrIO
MW: 310.9200 g/mol
MDL Number: MFCD07779026
CAS Number: 689291-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR