5-Chloro-3-hydroxypyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Chloro-5-pyridinol, 3-Chloro-5-hydroxypyridine, 5-Chloro-3-pyridinol

073219-1G 073219-5G 073219-10G
102529-012EA 107.69 USD
102529-012 102529-014 102529-016
5-Chloro-3-hydroxypyridine ≥95%
5-Chloro-3-hydroxypyridine

Hazard: Irritant

Formula: C₅H₄ClNO
MW: 129.55 g/mol
MDL Number: MFCD00006379
CAS Number: 74115-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR