2-Bromo-4-fluoro-5-nitrophenol ≥95%
Supplier: Matrix Scientific
073218-1G
073218-5G
073218-10G
102529-006EA
73.85
USD
102529-006
102529-008
102529-010
2-Bromo-4-fluoro-5-nitrophenol ≥95%
2-Bromo-4-fluoro-5-nitrophenol
Hazard: Irritant
Formula:
C₆H₃BrFNO₃ MW: 236.00 g/mol |
MDL Number:
MFCD08460104 CAS Number: 84478-87-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...