2,4-Dichloro-6-(trifluoromethyl)pyrimidine ≥95%

Supplier: Matrix Scientific

073216-1G 073216-5G
102528-994EA 1249.23 USD
102528-994 102528-996
2,4-Dichloro-6-(trifluoromethyl)pyrimidine ≥95%
2,4-Dichloro-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₅HCl₂F₃N₂
MW: 216.97 g/mol
MDL Number: MFCD09910264
CAS Number: 16097-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR