2,4,6-Trichloro-5-methylpyrimidine ≥95%

Supplier: Matrix Scientific

073214-1G 073214-5G 073214-10G
102528-982EA 53.85 USD
102528-982 102528-984 102528-986
2,4,6-Trichloro-5-methylpyrimidine ≥95%
2,4,6-Trichloro-5-methylpyrimidine

Hazard: Irritant

Formula: C₅H₃Cl₃N₂
MDL Number: MFCD00233525
CAS Number: 1780-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR