N-Allyl-N-(2-(5-methoxy-1H-indol-3-yl)ethyl)prop-2-en-1-amine ≥95%
Supplier: Matrix Scientific
073209-1G
073209-5G
102528-954EA
427.69
USD
102528-954
102528-956
N-Allyl-N-(2-(5-methoxy-1H-indol-3-yl)ethyl)prop-2-en-1-amine ≥95%
N-Allyl-N-(2-(5-methoxy-1H-indol-3-yl)ethyl)prop-2-en-1-amine
Hazard: Irritant
Formula:
C₁₇H₂₂N₂O |
CAS Number:
928822-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...