1-Acetyl-4-(methylamino)piperidine ≥95%

Supplier: Matrix Scientific

073208-1G 073208-5G
102528-950EA 640 USD
102528-950 102528-952
1-Acetyl-4-(methylamino)piperidine ≥95%
1-Acetyl-4-(methylamino)piperidine

Hazard: Irritant

Formula: C₈H₁₆N₂O
MW: 156.23 g/mol
MDL Number: MFCD07367773
CAS Number: 139062-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR