2-Chloroquinazoline ≥95%
Supplier: Matrix Scientific
073207-1G
073207-5G
102528-946EA
113.85
USD
102528-946
102528-948
2-Chloroquinazoline ≥95%
2-Chloroquinazoline
Hazard: Irritant
Formula:
C₈H₅ClN₂ MW: 164.5900 g/mol |
MDL Number:
MFCD09959715 CAS Number: 6141-13-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...