Methyl-3-amino-4-methylthiophene-2-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-amino-4-methyl-2-thiophenecarboxylate, 2-Thiophenecarboxylic acid, 3-amino-4-methyl-, methyl ester, Methyl 3-amino-4-methylthiophene-2-carboxylate

073206-10G 073206-100G
102528-940EA 49.23 USD
102528-940 102528-942
Methyl-3-amino-4-methylthiophene-2-carboxylate ≥95%
Methyl-3-amino-4-methylthiophene-2-carboxylate

Hazard: Irritant

Formula: C₇H₉NO₂S
MDL Number: MFCD00051822
CAS Number: 85006-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR