2,6-Dichloroimidazo[1,2-b]pyridazine ≥95%
Supplier: Matrix Scientific
073204-250MG
073204-1G
073204-5G
102528-928EA
498.46
USD
102528-928
102528-930
102528-932
2,6-Dichloroimidazo[1,2-b]pyridazine ≥95%
2,6-Dichloroimidazo[1,2-b]pyridazine
Hazard: Irritant
Formula:
C₆H₃Cl₂N₃ |
CAS Number:
112581-77-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...