2-Bromo-6-methoxypyrimidine ≥95%

Supplier: Matrix Scientific

073203-250MG 073203-1G 073203-5G
102528-922EA 203.56 USD
102528-922 102528-924 102528-926
2-Bromo-6-methoxypyrimidine ≥95%
2-Bromo-6-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₅BrN₂O
MW: 189.0100 g/mol
MDL Number: MFCD09702031
CAS Number: 944709-74-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR