4-Bromo-7-chloroquinoline ≥95%

Supplier: Matrix Scientific

073200-1G 073200-5G
102528-908EA 415.38 USD
102528-908 102528-910
4-Bromo-7-chloroquinoline ≥95%
4-Bromo-7-chloroquinoline

Hazard: Irritant

Formula: C₉H₅BrClN
MW: 242.5000 g/mol
MDL Number: MFCD07700232
CAS Number: 98519-65-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR