7-Benzenesulfonyl-4-chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%

Supplier: Matrix Scientific

073199-250MG 073199-1G
102528-904EA 967.69 USD
102528-904 102528-906
7-Benzenesulfonyl-4-chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine ≥95%
7-Benzenesulfonyl-4-chloro-6-methyl-7H-pyrrolo[2,3-d]pyrimidine

Hazard: Irritant

Formula: C₁₃H₁₀ClN₃O₂S
CAS Number: 252723-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR