5-(Aminomethyl)-2-bromopyridine ≥95%

Supplier: Matrix Scientific

073198-1G 073198-5G
102528-900EA 423.08 USD
102528-900 102528-902
5-(Aminomethyl)-2-bromopyridine ≥95%
5-(Aminomethyl)-2-bromopyridine

Hazard: Irritant

Formula: C₆H₇BrN₂
MDL Number: MFCD06738744
CAS Number: 120740-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR