5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 2-bromo-6,7-dihydro[1,3]thiazolo[5,4-c]pyridine-5(4H)-carboxylate, tert-Butyl 2-bromo-6,7-dihydrothiazolo[5,4-c]pyridine-5(4H)-carboxylate

073196-250MG 073196-1G
102528-890EA 86.15 USD
102528-890 102528-892
5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine ≥95%
5-Boc-2-bromo-6,7-dihydro-4H-thiazolo[5,4-c]pyridine

Hazard: Irritant

Formula: C₁₁H₁₅BrN₂O₂S
MW: 319.23 g/mol
MDL Number: MFCD09951958
CAS Number: 365996-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR