6-Bromo-1H-pyrrolo[2,3-b]pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: 6-Bromo-7-azaindole

073194-250MG 073194-1G
102528-880EA 247.69 USD
102528-880 102528-882
6-Bromo-1H-pyrrolo[2,3-b]pyridine ≥95%
6-Bromo-1H-pyrrolo[2,3-b]pyridine

Hazard: Irritant

Formula: C₇H₅BrN₂
MW: 197.04 g/mol
MDL Number: MFCD06659667
CAS Number: 143468-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR