tert-Butyl-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

073193-1G 073193-5G
102528-876EA 710.77 USD
102528-876 102528-878
tert-Butyl-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate ≥95%
tert-Butyl-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₇NO₂S
MW: 239.3300 g/mol
MDL Number: MFCD11518948
CAS Number: 230301-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR