Pyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione ≥95%

Supplier: Matrix Scientific

073192-250MG 073192-1G 073192-5G
102528-870EA 640 USD
102528-870 102528-872 102528-874
Pyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione ≥95%
Pyrido[3,4-d]pyrimidine-2,4(1H,3H)-dione

Hazard: Irritant

Formula: C7H5N3O2
MW: 163.1335 g/mol
MDL Number: MFCD07438024
CAS Number: 21038-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR