4-Chloro-8-methoxyquinazoline ≥95%

Supplier: Matrix Scientific

073190-250MG 073190-1G
102528-860EA 498.46 USD
102528-860 102528-862
4-Chloro-8-methoxyquinazoline ≥95%
4-Chloro-8-methoxyquinazoline

Hazard: Irritant

Formula: C₉H₇ClN₂O
MW: 194.6200 g/mol
MDL Number: MFCD09954869
CAS Number: 154288-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR