4-Bromoquinoline-6-carboxylic acid ≥95%

Supplier: Matrix Scientific

073188-250MG 073188-1G 073188-5G
102528-850EA 400 USD
102528-850 102528-852 102528-854
4-Bromoquinoline-6-carboxylic acid ≥95%
4-Bromoquinoline-6-carboxylic acid

Hazard: Irritant

Formula: C₁₀H₆BrNO₂
MW: 252.0600 g/mol
MDL Number: MFCD11518955
CAS Number: 219763-87-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR