4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine ≥95%

Supplier: Matrix Scientific

073187-250MG 073187-1G 073187-5G
102528-844EA 967.69 USD
102528-844 102528-846 102528-848
4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine ≥95%
4,6-Dichloro-1H-pyrrolo[3,2-c]pyridine

Hazard: Irritant

Formula: C₇H₄Cl₂N₂
MDL Number: MFCD09038480
CAS Number: 67139-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR