7-(Trifluoromethyl)quinoline-3-carboxylic acid ≥95%

Supplier: Matrix Scientific

073186-250MG 073186-1G
102528-840EA 684.62 USD
102528-840 102528-842
7-(Trifluoromethyl)quinoline-3-carboxylic acid ≥95%
7-(Trifluoromethyl)quinoline-3-carboxylic acid

Hazard: Irritant

Formula: C₁₁H₆F₃NO₂
MW: 241.1700 g/mol
MDL Number: MFCD11042923
CAS Number: 71082-51-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR