6,7-Dihydro-5H-pyrrolo[3,4-b]pyridine dihydrochloride ≥95%

Supplier: Matrix Scientific

073185-1G 073185-5G 073185-10G
102528-834EA 640 USD
102528-834 102528-836 102528-838
6,7-Dihydro-5H-pyrrolo[3,4-b]pyridine dihydrochloride ≥95%
6,7-Dihydro-5H-pyrrolo[3,4-b]pyridine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₀Cl₂N₂
MDL Number: MFCD09607967
CAS Number: 147740-02-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR